ENA296275062 Enamine

2-(1,3-dioxaindan-5-yl)-4-(chloromethyl)-1,3-thiazole

  • MDL number MFCD04054556
  • Cas Number : 55315-33-8
  • Molecular Weight : 253.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1,3-benzodioxol-5-yl)-4-(chloromethyl)-1,3-thiazole
Inchi Code 1S/C11H8ClNO2S/c12-4-8-5-16-11(13-8)7-1-2-9-10(3-7)15-6-14-9/h1-3,5H,4,6H2
Inchi Key DXTWZFZBGJRQHR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65085
Availability details for
Quantity
  • Availability
  • Add to Cart