ENA296275066 Enamine

quinoline-5-sulfonamide

  • MDL number MFCD02752662
  • Cas Number : 415913-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65094
Availability details for
Quantity
  • Availability
  • Add to Cart