ENA296275067 Enamine

5-(3-chlorophenyl)-4-methylthiophene-2-carboxylic acid

  • MDL number MFCD11189723
  • Cas Number : 1039859-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65096
Availability details for
Quantity
  • Availability
  • Add to Cart