ENA296275069 Enamine

(2S)-1-acetyl-2,3-dihydro-1H-indole-2-carboxylic acid

  • MDL number MFCD04972422
  • Cas Number : 82950-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-215
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65099
Availability details for
Quantity
  • Availability
  • Add to Cart