ENA296275072 Enamine

3-[5-(methylsulfanyl)-1H-1,2,3,4-tetrazol-1-yl]aniline

  • MDL number MFCD16622021
  • Cas Number : 1173146-92-3
  • Molecular Weight : 207.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-57756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57756
Availability details for
Quantity
  • Availability
  • Add to Cart