ENA296275078 Enamine

3-(ethoxymethyl)aniline

  • MDL number MFCD06804495
  • Cas Number : 80171-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65110
Availability details for
Quantity
  • Availability
  • Add to Cart