ENA296275079 Enamine

2-methylpropane-1,2-diol

  • MDL number MFCD00971926
  • Cas Number : 558-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65111

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-65111
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart