ENA296275080 Enamine

[2-chloro-6-(dimethylamino)phenyl]methanol

  • MDL number MFCD10695096
  • Cas Number : 1152515-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65112
Availability details for
Quantity
  • Availability
  • Add to Cart