ENA296275088 Enamine

5-(4-iodophenyl)pentanoic acid

  • MDL number MFCD11188214
  • Cas Number : 116680-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65121

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65121
Availability details for
Quantity
  • Availability
  • Add to Cart