ENA296275090 Enamine

2-(3-chlorobenzenesulfonamido)-2-phenylacetic acid

  • MDL number MFCD13358808
  • Cas Number : 1132880-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65124
Availability details for
Quantity
  • Availability
  • Add to Cart