ENA296275092 Enamine

2-(4-acetylbenzenesulfonamido)-3-carbamoylpropanoic acid

  • MDL number MFCD13354925
  • Cas Number : 1396962-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65126
Availability details for
Quantity
  • Availability
  • Add to Cart