ENA296275093 Enamine

3-carbamoyl-2-(3,4-dimethylbenzenesulfonamido)propanoic acid

  • MDL number MFCD13357477
  • Cas Number : 1396962-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65127
Availability details for
Quantity
  • Availability
  • Add to Cart