ENA296275095 Enamine

4-fluoro-2-(4-methoxybenzamido)benzoic acid

  • MDL number MFCD09732689
  • Cas Number : 953897-72-8
  • Molecular Weight : 289.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65131
Availability details for
Quantity
  • Availability
  • Add to Cart