ENA296275097 Enamine

5-(methoxymethyl)-2-methylfuran-3-carboxylic acid

  • MDL number MFCD13679827
  • Cas Number : 79347-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23529
Availability details for
Quantity
  • Availability
  • Add to Cart