ENA296275098 Enamine

2-(2,5-difluorophenyl)-2-(methylamino)acetic acid

  • MDL number MFCD12047753
  • Cas Number : 1192660-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65134
Availability details for
Quantity
  • Availability
  • Add to Cart