ENA296275099 Enamine

2-amino-1-(1,2,3,4-tetrahydroisoquinolin-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD13561989
  • Cas Number : 1220036-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65135

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65135
Availability details for
Quantity
  • Availability
  • Add to Cart