ENA296275103 Enamine

3-[(2-chlorophenoxy)methyl]-1-benzofuran-2-carboxylic acid

  • MDL number MFCD13381158
  • Cas Number : 1039891-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65139
Availability details for
Quantity
  • Availability
  • Add to Cart