ENA296275105 Enamine

1-(2-aminophenyl)-5-methyl-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD12913287
  • Cas Number : 1208528-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60389
Availability details for
Quantity
  • Availability
  • Add to Cart