ENA296275106 Enamine

1,1,1-triethoxybutane

  • MDL number MFCD00059382
  • Cas Number : 24964-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60547

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60547
Availability details for
Quantity
  • Availability
  • Add to Cart