ENA296275107 Enamine

N-(3-isothiocyanatopropyl)-N-methylaniline

  • MDL number MFCD11131109
  • Cas Number : 53485-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65155
Availability details for
Quantity
  • Availability
  • Add to Cart