ENA296275110 Enamine

3-[(2,2,2-trifluoroethoxy)methyl]aniline

  • MDL number MFCD09811898
  • Cas Number : 117401-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65158
Availability details for
Quantity
  • Availability
  • Add to Cart