ENA296275116 Enamine

(5-phenyl-1,3,4-oxadiazol-2-yl)methanol

  • MDL number MFCD01755392
  • Cas Number : 54014-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65170
Availability details for
Quantity
  • Availability
  • Add to Cart