ENA296275117 Enamine

1-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}propan-1-amine

  • MDL number MFCD11108300
  • Cas Number : 1092287-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65175
Availability details for
Quantity
  • Availability
  • Add to Cart