ENA296275126 Enamine

3-(difluoromethoxy)-2-methylaniline

  • MDL number MFCD12170011
  • Cas Number : 1154695-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65187
Availability details for
Quantity
  • Availability
  • Add to Cart