ENA296275130 Enamine

2-amino-N,6-dimethylbenzamide

  • MDL number MFCD09731040
  • Cas Number : 953727-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65193
Availability details for
Quantity
  • Availability
  • Add to Cart