ENA296275135 Enamine

2,3-dihydro-1H-indene-5-sulfonamide

  • MDL number MFCD00021245
  • Cas Number : 35203-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27000

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27000
Availability details for
Quantity
  • Availability
  • Add to Cart