ENA296275137 Enamine

3-(3-fluorophenyl)-1-[(piperidin-3-yl)methyl]urea

  • MDL number MFCD12862412
  • Cas Number : 1183803-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65202
Availability details for
Quantity
  • Availability
  • Add to Cart