ENA296275143 Enamine

N-(2-hydroxyethyl)methanesulfonamide

  • MDL number MFCD11611046
  • Cas Number : 6601-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65217
Availability details for
Quantity
  • Availability
  • Add to Cart