ENA296275145 Enamine

1-(1-benzofuran-2-ylmethyl)piperazine

  • MDL number MFCD12780500
  • Cas Number : 87813-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65221

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65221
Availability details for
Quantity
  • Availability
  • Add to Cart