ENA296275146 Enamine

1-(4-bromo-2-fluorophenyl)-4,4-dimethylpentan-3-one

  • MDL number MFCD12170236
  • Cas Number : 1156853-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65222
Availability details for
Quantity
  • Availability
  • Add to Cart