ENA296275147 Enamine

3-(4-bromophenyl)-3-methylpyrrolidine-2,5-dione

  • MDL number MFCD11207821
  • Cas Number : 1094289-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65223
Availability details for
Quantity
  • Availability
  • Add to Cart