ENA296275152 Enamine

2-(3-amino-1H-1,2,4-triazol-1-yl)-1-(azepan-1-yl)ethan-1-one

  • MDL number MFCD12766706
  • Cas Number : 1183925-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65232
Availability details for
Quantity
  • Availability
  • Add to Cart