ENA296275154 Enamine

2-amino-1-(piperazin-1-yl)ethan-1-one dihydrochloride

  • MDL number MFCD16622104
  • Cas Number : 1219158-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65234
Availability details for
Quantity
  • Availability
  • Add to Cart