ENA296275160 Enamine

3,3-bis(methylsulfanyl)-1-(pyridin-3-yl)prop-2-en-1-one

  • MDL number MFCD16622106
  • Cas Number : 68192-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65242
Availability details for
Quantity
  • Availability
  • Add to Cart