ENA296275165 Enamine

cinnoline-3-carboxylic acid

  • MDL number MFCD09383889
  • Cas Number : 90418-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-206
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27679
Availability details for
Quantity
  • Availability
  • Add to Cart