ENA296275171 Enamine

1-cyclopentyl-5-methanesulfonyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD11105970
  • Cas Number : 1152878-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65262
Availability details for
Quantity
  • Availability
  • Add to Cart