ENA296275173 Enamine

1,4-diethoxy-2-isothiocyanatobenzene

  • MDL number MFCD11124608
  • Cas Number : 99855-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65264
Availability details for
Quantity
  • Availability
  • Add to Cart