ENA296275174 Enamine

3-methyl-1-[3-(trifluoromethyl)phenyl]butan-1-amine

  • MDL number MFCD11148423
  • Cas Number : 1020972-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65265
Availability details for
Quantity
  • Availability
  • Add to Cart