ENA296275186 Enamine

N-benzyl-1,3-thiazol-2-amine

  • MDL number MFCD02131295
  • Cas Number : 41593-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65278
Availability details for
Quantity
  • Availability
  • Add to Cart