ENA296275192 Enamine

1-(5-chloro-2-methoxyphenyl)pyrrolidin-3-amine

  • MDL number MFCD11894833
  • Cas Number : 1179733-71-1
  • Molecular Weight : 226.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65295
Availability details for
Quantity
  • Availability
  • Add to Cart