ENA296275193 Enamine

(6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl)methanamine hydrochloride

  • MDL number MFCD07772790
  • Cas Number : 859833-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65296
Availability details for
Quantity
  • Availability
  • Add to Cart