ENA296275197 Enamine

thian-3-one

  • MDL number MFCD00040796
  • Cas Number : 19090-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62419

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-62419
pictograms GHS02
hstatements H225
Availability details for
Quantity
  • Availability
  • Add to Cart