ENA296275201 Enamine

3-(dodecyloxy)propanenitrile

  • MDL number MFCD00019869
  • Cas Number : 56637-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65311
Availability details for
Quantity
  • Availability
  • Add to Cart