ENA296275205 Enamine

3-methyl-1-[3-(trifluoromethyl)phenyl]butan-1-one

  • MDL number MFCD12153253
  • Cas Number : 1184224-97-2
  • Molecular Weight : 230.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65316
Availability details for
Quantity
  • Availability
  • Add to Cart