ENA296275207 Enamine

1-{3'-methyl-[1,1'-biphenyl]-3-yl}methanamine

  • MDL number MFCD05978949
  • Cas Number : 1176552-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65318
Availability details for
Quantity
  • Availability
  • Add to Cart