ENA296275210 Enamine

8,9-dichloro-2,3,4,5-tetrahydro-1-benzoxepin-5-amine

  • MDL number MFCD11104784
  • Cas Number : 1152598-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65321
Availability details for
Quantity
  • Availability
  • Add to Cart