ENA296275212 Enamine

2-{[4-(dimethylamino)phenyl]formamido}propanoic acid

  • MDL number MFCD09951129
  • Cas Number : 1396965-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65323
Availability details for
Quantity
  • Availability
  • Add to Cart