ENA296275213 Enamine

2-chloro-5-[4-(trifluoromethyl)phenoxymethyl]pyridine

  • MDL number MFCD12558745
  • Cas Number : 1039969-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65324

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65324
Availability details for
Quantity
  • Availability
  • Add to Cart