ENA296275215 Enamine

(4-difluoromethanesulfonylphenyl)hydrazine

  • MDL number MFCD12174238
  • Cas Number : 4837-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65327
Availability details for
Quantity
  • Availability
  • Add to Cart