ENA296275217 Enamine

2-(chloromethyl)-5-(2,6-dichlorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD09815570
  • Cas Number : 1016503-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65330
Availability details for
Quantity
  • Availability
  • Add to Cart